4-Bromo-2-chloro-6-iodoanisole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1-chloro-3-iodo-2-methoxybenzene

OR400944-1G OR400944-250MG OR400944-5G
APOSOR400944-1GEA 522 EUR
APOSOR400944-1G APOSOR400944-250MG APOSOR400944-5G
4-Bromo-2-chloro-6-iodoanisole 95%
4-Bromo-2-chloro-6-iodoanisole
Formel: C₇H₅BrClIO
Molecular Weight: 347,375 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31561245
CAS-Nummer: 861083-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR