4-Chloro-3-methoxybenzyl alcohol

Lieferant: Apollo Scientific

Synonyme: (4-Chloro-3-methoxyphenyl)methanol

OR400954-1G OR400954-5G
APOSOR400954-1GEA 89.9 EUR
APOSOR400954-1G APOSOR400954-5G
4-Chloro-3-methoxybenzyl alcohol
4-Chloro-3-methoxybenzyl alcohol
Formel: C₈H₉ClO₂
Molecular Weight: 172.609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527076
CAS-Nummer: 13726-17-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR