3-Chloro-4-iodobenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

OR400978-1G OR400978-250MG
APOSOR400978-1GEA 140 EUR
APOSOR400978-1G APOSOR400978-250MG
3-Chloro-4-iodobenzyl alcohol 95%
3-Chloro-4-iodobenzyl alcohol
Formel: C₇H₆ClIO
Molecular Weight: 268.479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393355
CAS-Nummer: 166386-60-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR