3-Chloro-4-iodobenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR400985-100MG
OR400985-1G
OR400985-250MG
APOSOR400985-100MGEA
21.9
EUR
APOSOR400985-100MG
APOSOR400985-1G
APOSOR400985-250MG
3-Chloro-4-iodobenzonitrile
3-Chloro-4-iodobenzonitrile
Formel:
C₇H₃ClIN Molecular Weight: 263,463 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393347 CAS-Nummer: 1261686-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...