3-Chloro-4-methoxybenzyl bromide

Lieferant: Apollo Scientific

Synonyme: 4-(Bromomethyl)-2-chloro-1-methoxybenzene

OR400991-5G
APOSOR400991-5GEA 171 EUR
APOSOR400991-5G
3-Chloro-4-methoxybenzyl bromide
3-Chloro-4-methoxybenzyl bromide
Formel: C₈H₈BrClO
Molecular Weight: 235,506 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06858766
CAS-Nummer: 320407-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR