3-Chloro-2-nitrobenzonitrile
Lieferant: Apollo Scientific
OR401015-5G
APOSOR401015-5GEA
133
EUR
APOSOR401015-5G
3-Chloro-2-nitrobenzonitrile
3-Chloro-2-nitrobenzonitrile
Formel:
C₇H₃ClN₂O₂ Molecular Weight: 182,564 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15529744 CAS-Nummer: 34662-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...