Methyl 3-chloro-2-nitrobenzoate

Lieferant: Apollo Scientific

OR401016-1G OR401016-5G
APOSOR401016-1GEA 58 EUR
APOSOR401016-1G APOSOR401016-5G
Methyl 3-chloro-2-nitrobenzoate
Methyl 3-chloro-2-nitrobenzoate
Formel: C₈H₆ClNO₄
Molecular Weight: 215.591 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038813
CAS-Nummer: 42087-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR