Methyl 3-Bromo-5-(bromomethyl)benzoate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-bromo-5-(bromomethyl)benzoate

OR401020-1G OR401020-250MG
APOSOR401020-1GEA 102 EUR
APOSOR401020-1G APOSOR401020-250MG
Methyl 3-Bromo-5-(bromomethyl)benzoate 98%
Methyl 3-Bromo-5-(bromomethyl)benzoate
Formel: C₉H₈Br₂O₂
Molecular Weight: 307.967 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044991
CAS-Nummer: 877624-40-3
UN: 3261
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR