4-Iodo-2-methoxybenzaldehyde

Lieferant: Apollo Scientific
Gefahr

OR401047-1G OR401047-250MG OR401047-5G
APOSOR401047-1GEA 288 EUR
APOSOR401047-1G APOSOR401047-250MG APOSOR401047-5G
4-Iodo-2-methoxybenzaldehyde
4-Iodo-2-methoxybenzaldehyde
Formel: C₈H₇IO₂
Molecular Weight: 262,044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20924440
CAS-Nummer: 139102-37-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR