2-Iodo-3-nitrobenzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR401052-1G
OR401052-250MG
OR401052-5G
APOSOR401052-1GEA
0
EUR
APOSOR401052-1G
APOSOR401052-250MG
APOSOR401052-5G
2-Iodo-3-nitrobenzaldehyde 95%
2-Iodo-3-nitrobenzaldehyde
Formel:
C₇H₄INO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18397935 CAS-Nummer: 1261481-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...