2-Iodo-3-nitrobenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

OR401053-1G OR401053-250MG OR401053-5G
APOSOR401053-1GEA 522 EUR
APOSOR401053-1G APOSOR401053-250MG APOSOR401053-5G
2-Iodo-3-nitrobenzyl alcohol 95%
2-Iodo-3-nitrobenzyl alcohol
Formel: C₇H₆INO₃
Molecular Weight: 279,032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18397984
CAS-Nummer: 158616-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR