3-Iodo-5-nitrobenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Iodo-5-nitrophenyl)methanol

OR401062-1G OR401062-250MG OR401062-5G
APOSOR401062-1GEA 33.5 EUR
APOSOR401062-1G APOSOR401062-250MG APOSOR401062-5G
3-Iodo-5-nitrobenzyl alcohol 95%
3-Iodo-5-nitrobenzyl alcohol
Formel: C₇H₆INO₃
Molecular Weight: 279,032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18397990
CAS-Nummer: 219589-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR