3-Iodo-5-nitrobenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR401063-1G
OR401063-250MG
OR401063-5G
APOSOR401063-1GEA
34.8
EUR
APOSOR401063-1G
APOSOR401063-250MG
APOSOR401063-5G
3-Iodo-5-nitrobenzyl bromide 95%
3-Iodo-5-nitrobenzyl bromide
Formel:
C₇H₅BrINO₂ Molecular Weight: 341,929 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18398007 CAS-Nummer: 1261644-60-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...