4-Acetyl-2-methylbenzonitrile

Lieferant: Apollo Scientific

Synonyme: 4'-Cyano-3'-methylacetophenone, 2-Cyano-5-acetyltoluene

OR40110-1G OR40110-5G
APOSOR40110-1GEA 522 EUR
APOSOR40110-1G APOSOR40110-5G
4-Acetyl-2-methylbenzonitrile
4-Acetyl-2-methylbenzonitrile
Formel: C₁₀H₉NO
Molecular Weight: 159,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845792
CAS-Nummer: 1138444-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR