6-Iodoisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR40112-1G
OR40112-250MG
APOSOR40112-1GEA
562
EUR
APOSOR40112-1G
APOSOR40112-250MG
6-Iodoisoquinoline
6-Iodoisoquinoline
Formel:
C₉H₆IN Molecular Weight: 255,055 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109663 CAS-Nummer: 75476-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...