4-Bromo-7-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR40120-250MG OR40120-500MG
APOSOR40120-250MGEA 139 EUR
APOSOR40120-250MG APOSOR40120-500MG
4-Bromo-7-methyl-1H-indazole
4-Bromo-7-methyl-1H-indazole
Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12028617
CAS-Nummer: 1159511-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR