5-Bromo-2,6-dimethyl-2H-indazole 95+%

Lieferant: Apollo Scientific
Achtung

OR40140-250MG OR40140-100MG
APOSOR40140-250MGEA 75.4 EUR
APOSOR40140-250MG APOSOR40140-100MG
5-Bromo-2,6-dimethyl-2H-indazole 95+%
5-Bromo-2,6-dimethyl-2H-indazole
Formel: C₉H₉BrN₂
Lagertemperatur: Kühlschrank
MDL: MFCD12028636
CAS-Nummer: 1159511-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR