5-Methyl-1H-indazole-6-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Methyl-1H-indazol-6-yl)boronic acid

OR40156-1G OR40156-250MG
APOSOR40156-1GEA 78.3 EUR
APOSOR40156-1G APOSOR40156-250MG
5-Methyl-1H-indazole-6-boronic acid
5-Methyl-1H-indazole-6-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175,98 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12028648
CAS-Nummer: 1310383-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR