1,5-Dimethyl-1H-indazole-6-boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Borono-1,5-dimethyl-1H-indazole
OR40167-1G
OR40167-250MG
APOSOR40167-1GEA
136
EUR
APOSOR40167-1G
APOSOR40167-250MG
1,5-Dimethyl-1H-indazole-6-boronic acid
1,5-Dimethyl-1H-indazole-6-boronic acid
Formel:
C₉H₁₁BN₂O₂ Molecular Weight: 190.007 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12028659 CAS-Nummer: 1310383-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...