5-Bromo-3-chloroisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR40227-1G
OR40227-250MG
APOSOR40227-1GEA
215
EUR
APOSOR40227-1G
APOSOR40227-250MG
5-Bromo-3-chloroisoquinoline
5-Bromo-3-chloroisoquinoline
Formel:
C₉H₅BrClN Molecular Weight: 242,5 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11044700 CAS-Nummer: 1029720-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...