3-Amino-4-butoxybenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Butoxy-5-carboxyaniline
OR40322-1G
OR40322-250MG
OR40322-500MG
APOSOR40322-1GEA
306
EUR
APOSOR40322-1G
APOSOR40322-250MG
APOSOR40322-500MG
3-Amino-4-butoxybenzoic acid
3-Amino-4-butoxybenzoic acid
Formel:
C₁₁H₁₅NO₃ Molecular Weight: 209,242 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12168705 CAS-Nummer: 855202-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...