3-Amino-4-(isopentyloxy)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-4-(3-methylbutoxy)benzoic acid, 5-Carboxy-2-(isopentyloxy)aniline
OR40323-1G
OR40323-250MG
OR40323-500MG
APOSOR40323-1GEA
328
EUR
APOSOR40323-1G
APOSOR40323-250MG
APOSOR40323-500MG
3-Amino-4-(isopentyloxy)benzoic acid
3-Amino-4-(isopentyloxy)benzoic acid
Formel:
C₁₂H₁₇NO₃ Molecular Weight: 223,268 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12168704 CAS-Nummer: 1096879-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...