N-(2,3-Dimethylbenzyl)cyclopropylamine hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: N-Cyclopropyl-2,3-dimethylbenzylamine hydrochloride, 3-[(Cyclopropylamino)methyl]-o-xylene hydrochloride

OR40329-1G OR40329-250MG
APOSOR40329-1GEA 621 EUR
APOSOR40329-1G APOSOR40329-250MG
N-(2,3-Dimethylbenzyl)cyclopropylamine hydrochloride
N-(2,3-Dimethylbenzyl)cyclopropylamine hydrochloride
Formel: C₁₂H₁₇N·ClH
Molecular Weight: 211,731 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17168270
CAS-Nummer: 909702-86-9
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR