3-Hydrazinopyridazine hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(Pyridazin-3-yl)hydrazine hydrochloride
OR40349-1G
OR40349-5G
OR40349-250MG
OR40349-500MG
APOSOR40349-1GEA
301
EUR
APOSOR40349-1G
APOSOR40349-5G
APOSOR40349-250MG
APOSOR40349-500MG
3-Hydrazinopyridazine hydrochloride 95%
3-Hydrazinopyridazine hydrochloride
Formel:
C₄H₆N₄·ClH Molecular Weight: 146,578 g/mol Schmelzpunkt: 210…212 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD11226159 CAS-Nummer: 117043-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...