2-Bromo-4-(tert-butyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

OR40352-250MG OR40352-1G
APOSOR40352-250MGEA 97.3 EUR
APOSOR40352-250MG APOSOR40352-1G
2-Bromo-4-(tert-butyl)pyridine 97%
2-Bromo-4-(tert-butyl)pyridine
Formel: C₉H₁₂BrN
Molecular Weight: 214.102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223221
CAS-Nummer: 50488-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR