3-Chloro-6-methoxypyridine-2-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-6-methoxypicolinaldehyde, 3-Chloro-2-formyl-6-methoxypyridine
OR40365-1G
OR40365-5G
APOSOR40365-1GEA
203
EUR
APOSOR40365-1G
APOSOR40365-5G
3-Chloro-6-methoxypyridine-2-carboxaldehyde
3-Chloro-6-methoxypyridine-2-carboxaldehyde
Formel:
C₇H₆ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189327 CAS-Nummer: 1060810-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...