3-Chloro-6-methoxypyridine-2-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Carbamoyl-3-chloro-6-methoxypyridine, 3-Chloro-6-methoxypicolinamide
OR40366-1G
OR40366-250MG
APOSOR40366-1GEA
503
EUR
APOSOR40366-1G
APOSOR40366-250MG
3-Chloro-6-methoxypyridine-2-carboxamide
3-Chloro-6-methoxypyridine-2-carboxamide
Formel:
C₇H₇ClN₂O₂ Molecular Weight: 186,596 g/mol Schmelzpunkt: 172…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD17676111 CAS-Nummer: 1257535-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...