3-Chloro-6-methoxypyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-6-methoxypicolinohydrazide, 3-Chloro-6-methoxypyridine-2-carboxylic acid hydrazide

OR40368-1G OR40368-250MG
APOSOR40368-1GEA 313 EUR
APOSOR40368-1G APOSOR40368-250MG
3-Chloro-6-methoxypyridine-2-carbohydrazide
3-Chloro-6-methoxypyridine-2-carbohydrazide
Formel: C₇H₈ClN₃O₂
Molecular Weight: 201,61 g/mol
Siedepunkt: 141…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17676113
CAS-Nummer: 1257535-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR