5-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-3,4-dihydro-2-methyl-3-oxo-2H-1,4-benzoxazine
OR40373-1G
OR40373-250MG
APOSOR40373-1GEA
348
EUR
APOSOR40373-1G
APOSOR40373-250MG
5-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
5-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Formel:
C₉H₁₀N₂O₂ Molecular Weight: 178,188 g/mol Schmelzpunkt: 181…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11915246 CAS-Nummer: 1176714-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...