3-Bromo-5-methoxybenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-formylanisole, 5-Bromo-m-anisaldehyde

OR40400-1G OR40400-5G OR40400-10G
APOSOR40400-1GEA 24.8 EUR
APOSOR40400-1G APOSOR40400-5G APOSOR40400-10G
3-Bromo-5-methoxybenzaldehyde 95%
3-Bromo-5-methoxybenzaldehyde
Formel: C₈H₇BrO₂
Molecular Weight: 215.044 g/mol
Schmelzpunkt: 68…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797975
CAS-Nummer: 262450-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR