Methyl pent-2-enoate 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Methyl 3-ethylacrylate

OR40582-25G OR40582-5G
APOSOR40582-25GEA 82.8 EUR
APOSOR40582-25G APOSOR40582-5G
Methyl pent-2-enoate 97%
Methyl pent-2-enoate
Formel: C₆H₁₀O₂
Molecular Weight: 114.142 g/mol
Siedepunkt: 81…82 °C (45 mm Hg)
Lagertemperatur: Gefrierschrank
MDL: MFCD00137611
CAS-Nummer: 818-59-7
UN: 1993
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR