2-Bromo-4-iodobenzyl bromide 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: α,2-Dibromo-4-iodotoluene, 2-Bromo-1-(bromomethyl)-4-iodobenzene

OR40660-250MG OR40660-1G OR40660-5G
APOSOR40660-250MGEA 109 EUR
APOSOR40660-250MG APOSOR40660-1G APOSOR40660-5G
2-Bromo-4-iodobenzyl bromide 95+%
2-Bromo-4-iodobenzyl bromide
Formel: C₇H₅Br₂I
Molecular Weight: 375,827 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391667
CAS-Nummer: 1261776-07-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR