4-Bromo-2-iodobenzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: α,4-Dibromo-2-iodotoluene, 4-Bromo-1-(bromomethyl)-2-iodobenzene

OR40663-1G OR40663-250MG OR40663-5G
APOSOR40663-1GEA 66.7 EUR
APOSOR40663-1G APOSOR40663-250MG APOSOR40663-5G
4-Bromo-2-iodobenzyl bromide 97%
4-Bromo-2-iodobenzyl bromide
Formel: C₇H₅Br₂I
Molecular Weight: 375,827 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391672
CAS-Nummer: 885681-96-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR