3-Bromo-5-iodobenzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR40676-1G
OR40676-5G
APOSOR40676-1GEA
116
EUR
APOSOR40676-1G
APOSOR40676-5G
3-Bromo-5-iodobenzaldehyde
3-Bromo-5-iodobenzaldehyde
Formel:
C₇H₄BrIO Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779024 CAS-Nummer: 188813-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...