5-Chloro-6-iodopyridine-2,3-diamine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR40768-1G OR40768-250MG
APOSOR40768-1GEA 534 EUR
APOSOR40768-1G APOSOR40768-250MG
5-Chloro-6-iodopyridine-2,3-diamine hydrochloride 95%
5-Chloro-6-iodopyridine-2,3-diamine hydrochloride
Formel: C₅H₅ClIN₃·ClH
Molecular Weight: 305,932 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31812085
CAS-Nummer: 2367002-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR