7-Chloro-1-iodophenazine

Lieferant: Apollo Scientific
Achtung

OR40774-1G OR40774-250MG OR40774-5G
APOSOR40774-1GEA 53.8 EUR
APOSOR40774-1G APOSOR40774-250MG APOSOR40774-5G
7-Chloro-1-iodophenazine
7-Chloro-1-iodophenazine
Formel: C₁₂H₆ClIN₂
Molecular Weight: 340.547 g/mol
Schmelzpunkt: 166…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD31920066
CAS-Nummer: 2367002-64-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR