4-Bromo-3-cyclopropylaniline 95%

Lieferant: Apollo Scientific
Achtung

OR40779-1G OR40779-250MG OR40779-5G
APOSOR40779-1GEA 522 EUR
APOSOR40779-1G APOSOR40779-250MG APOSOR40779-5G
4-Bromo-3-cyclopropylaniline 95%
4-Bromo-3-cyclopropylaniline
Formel: C₉H₁₀BrN
Molecular Weight: 212,086 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD21100004
CAS-Nummer: 1353855-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR