3-Bromoisothiazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-1,2-thiazole
OR40801-1G
OR40801-250MG
APOSOR40801-1GEA
741
EUR
APOSOR40801-1G
APOSOR40801-250MG
3-Bromoisothiazole 95%
3-Bromoisothiazole
Formel:
C₃H₂BrNS Molecular Weight: 164.024 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07778378 CAS-Nummer: 55512-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...