Isothiazole-3-carbonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Thiazole-3-carbonitrile, 3-Cyanoisothiazole
OR40802-1G
OR40802-250MG
APOSOR40802-1GEA
509
EUR
APOSOR40802-1G
APOSOR40802-250MG
Isothiazole-3-carbonitrile 95%
Isothiazole-3-carbonitrile
Formel:
C₄H₂N₂S Molecular Weight: 110.137 g/mol Schmelzpunkt: 60…61 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD15529707 CAS-Nummer: 1452-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...