3-(1-Aminoethyl)-1-adamantanol hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR40811-1G OR40811-25G OR40811-5G
APOSOR40811-1GEA 33.5 EUR
APOSOR40811-1G APOSOR40811-25G APOSOR40811-5G
3-(1-Aminoethyl)-1-adamantanol hydrochloride 95%
3-(1-Aminoethyl)-1-adamantanol hydrochloride
Formel: C₁₂H₂₁NO·ClH
Molecular Weight: 231.762 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01825645
CAS-Nummer: 128487-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR