1-Bromo-2-prop-2-ynoxy-benzene 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR40821-1G
OR40821-250MG
OR40821-5G
APOSOR40821-1GEA
34.8
EUR
APOSOR40821-1G
APOSOR40821-250MG
APOSOR40821-5G
1-Bromo-2-prop-2-ynoxy-benzene 98%
1-Bromo-2-prop-2-ynoxy-benzene
Formel:
C₉H₇BrO Lagertemperatur: Kühlschrank |
MDL:
MFCD09934815 CAS-Nummer: 38770-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...