7-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine

Lieferant: Apollo Scientific

OR41148-1G OR41148-250MG
APOSOR41148-1GEA 169 EUR
APOSOR41148-1G APOSOR41148-250MG
7-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine
7-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine
Formel: C₇H₆BrN₃
Molecular Weight: 212,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827554
CAS-Nummer: 1190927-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR