(1S,2S)-(+)-2-Methylcyclopropane-1-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(1S,2S)-2-methylcyclopropane-1-carboxylic acid
OR41152-1G
OR41152-500MG
APOSOR41152-1GEA
1790
EUR
APOSOR41152-1G
APOSOR41152-500MG
(1S,2S)-(+)-2-Methylcyclopropane-1-carboxylic acid 95%
(1S,2S)-(+)-2-Methylcyclopropane-1-carboxylic acid
Formel:
C₅H₈O₂ Molecular Weight: 100,116 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD19228287 CAS-Nummer: 14590-52-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...