(4S)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected 95%
Lieferant: Apollo Scientific
Achtung
Synonyme: (4S)-3-(tert-Butoxycarbonyl)-4-methyl-1,2,3-oxathiazolidine 2,2-dioxide, tert-Butyl (4S)-4-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide, (S)-tert-Butyl 4-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide
OR41174-1G OR41174-5G OR41174-25G
APOSOR41174-1GEA 52.2 EUR
APOSOR41174-1G APOSOR41174-5G APOSOR41174-25G
(4S)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected 95%
(4S)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected
Formel: C₈H₁₅NO₅S
Molecular Weight: 237,273 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28122917
CAS-Nummer: 439948-91-1