6-Bromo-4-chloro-7-methylquinoline

Lieferant: Apollo Scientific
Achtung

OR42034-1G OR42034-250MG
APOSOR42034-1GEA 476 EUR
APOSOR42034-1G APOSOR42034-250MG
6-Bromo-4-chloro-7-methylquinoline
6-Bromo-4-chloro-7-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674991
CAS-Nummer: 1189106-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR