3-(4-Bromophenyl)prop-2-yn-1-ol 97%

Lieferant: Apollo Scientific
Achtung

OR42051-1G OR42051-250MG OR42051-5G
APOSOR42051-1GEA 79.9 EUR
APOSOR42051-1G APOSOR42051-250MG APOSOR42051-5G
3-(4-Bromophenyl)prop-2-yn-1-ol 97%
3-(4-Bromophenyl)prop-2-yn-1-ol
Formel: C₉H₇BrO
Molecular Weight: 211.055 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01250510
CAS-Nummer: 37614-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR