1-Bromo-4-(3-bromoprop-1-ynyl)benzene 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-4-(3-bromoprop-1-yn-1-yl)benzene
OR42054-1G
OR42054-250MG
OR42054-5G
APOSOR42054-1GEA
98.6
EUR
APOSOR42054-1G
APOSOR42054-250MG
APOSOR42054-5G
1-Bromo-4-(3-bromoprop-1-ynyl)benzene 95%
1-Bromo-4-(3-bromoprop-1-ynyl)benzene
Formel:
C₉H₆Br₂ Molecular Weight: 273,952 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD22482470 CAS-Nummer: 173019-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...