2-Cyclopropyl-4-iodoaniline
Lieferant: Apollo Scientific
|
Achtung
|
OR42059-1G
OR42059-250MG
APOSOR42059-1GEA
0
EUR
APOSOR42059-1G
APOSOR42059-250MG
2-Cyclopropyl-4-iodoaniline
2-Cyclopropyl-4-iodoaniline
Formel:
C₉H₁₀IN Lagertemperatur: Gefrierschrank |
MDL:
MFCD27938271 CAS-Nummer: 1704065-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...