Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR42104-1G OR42104-250MG OR42104-5G
APOSOR42104-1GEA 75.4 EUR
APOSOR42104-1G APOSOR42104-250MG APOSOR42104-5G
Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate
Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate
Formel: C₈H₇NO₂S
Molecular Weight: 181,212 g/mol
Schmelzpunkt: 127…128,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07772830
CAS-Nummer: 118465-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR