8-Bromo-3-hydroxyisoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Bromoisoquinolin-3(2H)-one, 8-Bromoisoquinolin-3-ol

OR42113-500MG OR42113-1G OR42113-250MG
APOSOR42113-500MGEA 66.7 EUR
APOSOR42113-500MG APOSOR42113-1G APOSOR42113-250MG
8-Bromo-3-hydroxyisoquinoline
8-Bromo-3-hydroxyisoquinoline
Formel: C₉H₆BrNO
Molecular Weight: 224,054 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11846498
CAS-Nummer: 608515-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR